KEY465201364 Key Organics / BIONET

1-[(4-methylphenyl)methyl]-8-(trifluoromethyl)-2,3,4,5-tetrahydro-1H-1,5-benzodiazepin-2-one

  • MDL number MFCD04126423
  • Cas Number : 866149-49-7
  • Molecular Weight : 334.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94152

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/9W-0879_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart