KEY465201369 Key Organics / BIONET

2-{4-[5-(4-chlorophenyl)-1H-pyrazol-3-yl]piperidin-1-yl}-3-nitropyridine

  • MDL number MFCD05668939
  • Cas Number : 1030856-25-7
  • Molecular Weight : 383.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94170

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/9X-0037_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart