KEY465201416 Key Organics / BIONET

1-({6-chloroimidazo[1,2-a]pyridin-3-yl}methyl)-4-phenylpiperidine-4-carbonitrile

  • MDL number MFCD08064776
  • Cas Number : 882748-17-6
  • Molecular Weight : 350.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94352

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/9Z-0829_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart