KEY465201437 Key Organics / BIONET

methyl 3'-methyl-[1,1'-biphenyl]-3-carboxylate

  • MDL number MFCD05979105
  • Cas Number : 128460-74-2
  • Molecular Weight : 226.275

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/650234

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-3198_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart