KEY465201439 Key Organics / BIONET

2-methyl-9H-fluoren-9-one

  • MDL number MFCD01151943
  • Cas Number : 2840-51-9
  • Molecular Weight : 194.233

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/650236

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-3200_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart