KEY465201459 Key Organics / BIONET

ethyl 3,5-dimethyl-1-[4-(propan-2-yl)phenyl]-1H-pyrazole-4-carboxylate

  • MDL number MFCD11501745
  • Cas Number : 1172362-54-7
  • Molecular Weight : 286.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94905

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BA-0853_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart