KEY465201471 Key Organics / BIONET

methyl 2-(4-methylbenzenesulfonamido)acetate

  • MDL number MFCD00558890
  • Molecular Weight : 243.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl {[(4-methylphenyl)sulfonyl]amino}acetate
Inchi Code 1S/C10H13NO4S/c1-8-3-5-9(6-4-8)16(13,14)11-7-10(12)15-2/h3-6,11H,7H2,1-2H3
Inchi Key GYQBPYXMRNEAKZ-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95427

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/CA-0008_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart