KEY465201501 Key Organics / BIONET

4-(piperidin-1-yl)butanoic acid hydrochloride

  • MDL number MFCD06801215
  • Cas Number : 5463-76-3
  • Molecular Weight : 207.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(1-piperidinyl)butanoic acid hydrochloride
Inchi Code 1S/C9H17NO2.ClH/c11-9(12)5-4-8-10-6-2-1-3-7-10;/h1-8H2,(H,11,12);1H
Inchi Key IZVIJYFRXDQEOR-UHFFFAOYSA-N

Description

Purity 85%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/650278

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/CS-4355_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart