KEY465201505 Key Organics / BIONET

methyl 4-hydroxybut-2-ynoate

  • MDL number MFCD24396522
  • Cas Number : 31555-05-2
  • Molecular Weight : 114.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/650282

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/CS-4395_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart