KEY465201517 Key Organics / BIONET

6-hydroxy-5-(prop-2-en-1-yl)-3,4-dihydropyrimidin-4-one

  • MDL number MFCD17019403
  • Cas Number : 16019-30-0
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-allyl-4,6-pyrimidinediol
Inchi Code 1S/C7H8N2O2/c1-2-3-5-6(10)8-4-9-7(5)11/h2,4H,1,3H2,(H2,8,9,10,11)
Inchi Key QWYRCXSPNDGEAQ-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96055

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/DC-0032_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart