KEY465201526 Key Organics / BIONET

N-(2-hydroxyethyl)-1H-indole-2-carboxamide

  • MDL number MFCD09430098
  • Cas Number : 144969-97-1
  • Molecular Weight : 204.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96300

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/EA-0819_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart