KEY465201546 Key Organics / BIONET

1-(4-tert-butylphenyl)imidazolidin-2-one

  • MDL number MFCD11501858
  • Cas Number : 1092346-57-0
  • Molecular Weight : 218.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96793

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/FA-0891_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart