KEY465201567 Key Organics / BIONET

3-(piperidin-4-yl)benzoic acid

  • MDL number MFCD05864627
  • Cas Number : 766508-67-2
  • Molecular Weight : 205.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(4-piperidinyl)benzoic acid
Inchi Code 1S/C12H15NO2/c14-12(15)11-3-1-2-10(8-11)9-4-6-13-7-5-9/h1-3,8-9,13H,4-7H2,(H,14,15)
Inchi Key ICCKWGCLHGIEQE-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/403919

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/FS-2030_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart