KEY465201583 Key Organics / BIONET

1-(2,3-dichlorophenyl)piperazine

  • MDL number MFCD01075181
  • Cas Number : 41202-77-1
  • Molecular Weight : 231.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97519

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GF-0124_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart