KEY465201702 Key Organics / BIONET

N'-(diphenylmethylidene)-2-(piperidin-1-yl)acetohydrazide

  • MDL number MFCD01569237
  • Cas Number : 478247-21-1
  • Molecular Weight : 321.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93150

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/9N-507S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart