KEY465201808 Key Organics / BIONET

1-[(4-tert-butylphenyl)methyl]-4-(furan-2-yl)-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD03012367
  • Cas Number : 439120-71-5
  • Molecular Weight : 332.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93529

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/9R-0209_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart