KEY465201845 Key Organics / BIONET

4-(2-ethylbutanoyl)-N-[(thiophen-2-yl)methyl]-1H-pyrrole-2-carboxamide

  • MDL number MFCD02571483
  • Cas Number : 478249-20-6
  • Molecular Weight : 304.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93555

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/9R-0238_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart