KEY465201873 Key Organics / BIONET

N-[(furan-2-yl)methyl]-4-(2-phenylacetyl)-1H-pyrrole-2-carboxamide

  • MDL number MFCD03012383
  • Cas Number : 439120-83-9
  • Molecular Weight : 308.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/93581

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/9R-0270_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart