KEY465201931 Key Organics / BIONET

1-(4-fluorophenyl)-1,3-dihydro-2-benzofuran-5-carbonitrile

  • MDL number MFCD06200812
  • Cas Number : 64169-67-1
  • Molecular Weight : 239.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 95 - 97
bp 375�t 760 mmHg
Iupac Name 1-(4-fluorophenyl)-1,3-dihydro-2-benzofuran-5-carbonitrile
Inchi Code 1S/C15H10FNO/c16-13-4-2-11(3-5-13)15-14-6-1-10(8-17)7-12(14)9-18-15/h1-7,15H,9H2
Inchi Key YXCRMKYHFFMNPT-UHFFFAOYSA-N

Description

Melting Point 95 - 97
Purity 90%
Physical Form Solid
Country of Origin GB
Boiling point 375�t 760 mmHg
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97878

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301 + P312 + P330,P302 + P352 + P312,P304 + P340 + P311,P305 + P351 + P338,P332 + P313,P337 + P313,P362,P402 + P404,P411,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/HF-0004_SDS.pdf
hcodes H302,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart