KEY465201988 Key Organics / BIONET

2-({2-[(2-formyl-4-nitrophenyl)sulfanyl]ethyl}sulfanyl)-5-nitrobenzaldehyde

  • MDL number MFCD00169218
  • Cas Number : 331460-03-8
  • Molecular Weight : 392.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/98230

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-0148_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart