KEY465202010 Key Organics / BIONET

4-(4-bromophenyl)-2-(cyclohexylamino)-4-oxobutanoic acid

  • MDL number MFCD00169323
  • Cas Number : 1031263-33-8
  • Molecular Weight : 354.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/98327

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-0262_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart