KEY465202050 Key Organics / BIONET

4-[(1E)-3-(4-fluorophenyl)-3-oxoprop-1-en-1-yl]phenyl furan-2-carboxylate

  • MDL number MFCD00169517
  • Cas Number : 298215-40-4
  • Molecular Weight : 336.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/98536

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-0473_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart