KEY465202069 Key Organics / BIONET

methyl 3-[(4-fluoro-3-nitrophenyl)carbamoyl]propanoate

  • MDL number MFCD00169593
  • Cas Number : 331461-03-1
  • Molecular Weight : 270.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/98613

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-0552_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart