KEY465202080 Key Organics / BIONET

4-[(1E)-3-[(4-chlorophenyl)sulfanyl]-3-oxoprop-1-en-1-yl]benzonitrile

  • MDL number MFCD00169694
  • Cas Number : 331461-60-0
  • Molecular Weight : 299.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/98704

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-0652_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart