KEY465202174 Key Organics / BIONET

4-(4-bromophenyl)-4-oxo-2-{[(pyridin-3-yl)methyl]amino}butanoic acid

  • MDL number MFCD00954935
  • Cas Number : 1042701-72-3
  • Molecular Weight : 363.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/99208

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-1207_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart