KEY465202200 Key Organics / BIONET

5-iodo-2-methylbenzonitrile

  • MDL number MFCD00128256
  • Cas Number : 52107-68-3
  • Molecular Weight : 243.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-iodo-2-methylbenzonitrile
Inchi Code 1S/C8H6IN/c1-6-2-3-8(9)4-7(6)5-10/h2-4H,1H3
Inchi Key QPGQEWLZKVSLPK-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/99391

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-1420_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart