KEY465202203 Key Organics / BIONET

2-[(4-bromophenyl)sulfanyl]-4-(4-fluorophenyl)-4-oxobutanoic acid

  • MDL number MFCD00955113
  • Cas Number : 301194-41-2
  • Molecular Weight : 383.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/99425

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-1458_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart