KEY465202217 Key Organics / BIONET

3-amino-2-(thiophen-2-yl)-3,4-dihydroquinazolin-4-one

  • MDL number MFCD01331067
  • Cas Number : 329080-15-1
  • Molecular Weight : 243.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/99508

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-1547_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart