KEY465202255 Key Organics / BIONET

5-(4-chlorophenyl)-3-cyclopropyl-1-phenyl-4,5-dihydro-1H-pyrazole

  • MDL number MFCD02675236
  • Cas Number : 303091-42-1
  • Molecular Weight : 296.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/99758

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-1822_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart