KEY465202260 Key Organics / BIONET

3-(4-chlorophenyl)-3-[(4-chlorophenyl)sulfanyl]-1-cyclopropylpropan-1-one

  • MDL number MFCD02676113
  • Cas Number : 329779-49-9
  • Molecular Weight : 351.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/99826

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-1889_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart