KEY465202309 Key Organics / BIONET

1-(2H-1,3-benzodioxol-5-yl)-3-[(4-ethoxyphenyl)amino]propan-1-one

  • MDL number MFCD03196744
  • Cas Number : 717827-75-3
  • Molecular Weight : 313.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/100205

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-2269_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart