KEY465202368 Key Organics / BIONET

1-(4-bromophenyl)-3-[(4-ethylphenyl)amino]propan-1-one

  • MDL number MFCD00423361
  • Cas Number : 423735-02-8
  • Molecular Weight : 332.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-bromophenyl)-3-(4-ethylanilino)-1-propanone
Inchi Code 1S/C17H18BrNO/c1-2-13-3-9-16(10-4-13)19-12-11-17(20)14-5-7-15(18)8-6-14/h3-10,19H,2,11-12H2,1H3
Inchi Key BPDBFCSMKSAANN-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/100360

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-2436_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart