KEY465202470 Key Organics / BIONET

octahydrocyclopenta[c]pyrrole hydrochloride

  • MDL number MFCD06411019
  • Cas Number : 112626-50-3
  • Molecular Weight : 147.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name octahydrocyclopenta[c]pyrrole hydrochloride
Inchi Code 1S/C7H13N.ClH/c1-2-6-4-8-5-7(6)3-1;/h6-8H,1-5H2;1H
Inchi Key HVZRRRPCVOJOLJ-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101928

Safety Information

signalword Warning
pcodes P232,P261,P264,P270,P271,P280,P301 + P310,P301 + P312 + P330,P302 + P352 + P312,P304 + P340 + P311,P305 + P351 + P338,P332 + P313,P337 + P313,P362,P402 + P404,P422,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/LF-0504_SDS.pdf
hcodes H302,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart