KEY465202500 Key Organics / BIONET

1-(4-fluorophenyl)piperazine dihydrochloride

  • MDL number MFCD00012765
  • Cas Number : 64090-19-3
  • Molecular Weight : 253.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 231 - 234
Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101968

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/LF-0544_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart