KEY465202742 Key Organics / BIONET

(5E)-5-[(4-fluorophenyl)methylidene]-2-sulfanyl-4,5-dihydro-1,3-thiazol-4-one

  • MDL number MFCD04969012
  • Cas Number : 402-93-7
  • Molecular Weight : 239.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (5E)-5-(4-fluorobenzylidene)-2-thioxo-1,3-thiazolidin-4-one
Inchi Code 1S/C10H6FNOS2/c11-7-3-1-6(2-4-7)5-8-9(13)12-10(14)15-8/h1-5H,(H,12,13,14)/b8-5+
Inchi Key BQOQSIQMDZUHKF-VMPITWQZSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/103237

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/MS-1464_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart