KEY465202756 Key Organics / BIONET

2-cyano-N-(2,4-difluorophenyl)acetamide

  • MDL number MFCD00116881
  • Cas Number : 1961-38-2
  • Molecular Weight : 196.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/103300

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/MS-1556_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart