KEY465202764 Key Organics / BIONET

1-(2-chlorophenyl)-3,3-bis(methylsulfanyl)prop-2-en-1-one

  • MDL number MFCD05669632
  • Cas Number : 56944-67-3
  • Molecular Weight : 258.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/103322

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/MS-1584_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart