KEY465202799 Key Organics / BIONET

1-methyl-1,3-diazinane-2,4,6-trione

  • MDL number MFCD00127915
  • Cas Number : 2565-47-1
  • Molecular Weight : 142.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,6-dihydroxy-3-methyl-4(3H)-pyrimidinone
Inchi Code 1S/C5H6N2O3/c1-7-4(9)2-3(8)6-5(7)10/h2H2,1H3,(H,6,8,10)
Inchi Key DCGGMHIZEAHUJL-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/103490

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/MS-1844_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart