KEY465202824 Key Organics / BIONET

4-methoxy-2-methylbenzonitrile

  • MDL number MFCD03425692
  • Cas Number : 21883-13-6
  • Molecular Weight : 147.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-methoxy-2-methylbenzonitrile
Inchi Code 1S/C9H9NO/c1-7-5-9(11-2)4-3-8(7)6-10/h3-5H,1-2H3
Inchi Key PXAQQUKODRZAOZ-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/103563

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/MS-1940_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart