KEY465202846 Key Organics / BIONET

2-{[5-(4-fluorobenzamido)-1,3,4-thiadiazol-2-yl]sulfanyl}acetic acid

  • MDL number MFCD02169484
  • Cas Number : 329695-55-8
  • Molecular Weight : 313.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/103645

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/MS-2048_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart