KEY465202854 Key Organics / BIONET

3-{[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]carbamoyl}propanoic acid

  • MDL number MFCD01232130
  • Cas Number : 797806-64-5
  • Molecular Weight : 269.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-oxo-4-{[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]amino}butanoic acid
Inchi Code 1S/C7H6F3N3O3S/c8-7(9,10)5-12-13-6(17-5)11-3(14)1-2-4(15)16/h1-2H2,(H,15,16)(H,11,13,14)
Inchi Key PTYFZPOMQNHYHW-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/103670

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/MS-2075_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart