KEY465202855 Key Organics / BIONET

4-(4-nitrophenyl)morpholine

  • MDL number MFCD00023317
  • Cas Number : 10389-51-2
  • Molecular Weight : 208.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(4-nitrophenyl)morpholine
Inchi Code 1S/C10H12N2O3/c13-12(14)10-3-1-9(2-4-10)11-5-7-15-8-6-11/h1-4H,5-8H2
Inchi Key IAJDSUYFELYZCS-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/103673

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/MS-2079_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart