KEY465202862 Key Organics / BIONET

6-nitro-4-oxo-1,4-dihydroquinoline-2-carboxylic acid

  • MDL number MFCD02169438
  • Cas Number : 52980-08-2
  • Molecular Weight : 234.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-hydroxy-6-nitro-2-quinolinecarboxylic acid
Inchi Code 1S/C10H6N2O5/c13-9-4-8(10(14)15)11-7-2-1-5(12(16)17)3-6(7)9/h1-4H,(H,11,13)(H,14,15)
Inchi Key KQFRZQHCZNAUOS-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/103693

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/MS-2111_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart