KEY465202868 Key Organics / BIONET

methyl 4-methyl-1,3-thiazole-5-carboxylate

  • MDL number MFCD00129998
  • Cas Number : 81569-44-0
  • Molecular Weight : 157.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-methyl-1,3-thiazole-5-carboxylate
Inchi Code 1S/C6H7NO2S/c1-4-5(6(8)9-2)10-3-7-4/h3H,1-2H3
Inchi Key RRCLLMUIJYXSGZ-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/103714

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/MS-2142_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart