KEY465202897 Key Organics / BIONET

(2E)-3-(4-oxo-4H-chromen-3-yl)prop-2-enoic acid

  • MDL number MFCD01548049
  • Cas Number : 42059-58-5
  • Molecular Weight : 216.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2E)-3-(4-oxo-4H-chromen-3-yl)-2-propenoic acid
Inchi Code 1S/C12H8O4/c13-11(14)6-5-8-7-16-10-4-2-1-3-9(10)12(8)15/h1-7H,(H,13,14)/b6-5+
Inchi Key JTKDMUFCCJDYLT-AATRIKPKSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/103801

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/MS-2277_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart