KEY465202929 Key Organics / BIONET

N-(3-chlorophenyl)-2-methyl-1-oxa-4-azaspiro[4.6]undecane-4-carboxamide

  • MDL number MFCD02667629
  • Cas Number : 633299-25-9
  • Molecular Weight : 322.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/103892

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/MS-2391_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart