KEY465202934 Key Organics / BIONET

(NZ)-4-(dimethylamino)-N-oxido-N-[(phenylcarbamoyl)methylidene]anilinium

  • MDL number MFCD29042761
  • Cas Number : 32331-63-8
  • Molecular Weight : 283.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. Store at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/650341

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404,P411
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/NS-00041_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart