KEY465202936 Key Organics / BIONET

methyl 2,3,4,7-tetrahydro-1H-isoindole-2-carboxylate

  • MDL number MFCD29042763
  • Cas Number : 1823183-69-2
  • Molecular Weight : 179.219

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/650347

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/NS-00047_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart