KEY465203017 Key Organics / BIONET

2-[2-(2-aminophenoxy)ethoxy]aniline

  • MDL number MFCD00087648
  • Cas Number : 52411-34-4
  • Molecular Weight : 244.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-[2-(2-aminophenoxy)ethoxy]aniline
Inchi Code 1S/C14H16N2O2/c15-11-5-1-3-7-13(11)17-9-10-18-14-8-4-2-6-12(14)16/h1-8H,9-10,15-16H2
Inchi Key PSDFQEVOCCOOET-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/104116

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/MS-2652_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart