KEY465203100 Key Organics / BIONET

thiophene-3-carbohydrazide

  • MDL number MFCD01310840
  • Cas Number : 39001-23-5
  • Molecular Weight : 142.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-thiophenecarbohydrazide
Inchi Code 1S/C5H6N2OS/c6-7-5(8)4-1-2-9-3-4/h1-3H,6H2,(H,7,8)
Inchi Key AMOWAJLALKAYKL-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/104306

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/MS-2874_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart